Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SOCS6
- Entrez Gene ID
-
9306
- Gene Name
- suppressor of cytokine signaling 6
- Gene Aliases
-
CIS-4, CIS4, HSPC060, SOCS-4, SOCS-6, SOCS4, SSI4, STAI4, STATI4
- Chromosome Location
-
Chr.18: 70288901 - 70330199 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_004232.3 |
NP_004223.2 |
1 - 2 |
186 |
127 |
View Full Product Details