Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR483
- Entrez Gene ID
-
619552
- Gene Name
- microRNA 483
- Gene Aliases
-
MIRN483, hsa-mir-483, mir-483
- Chromosome Location
-
Chr.11: 2134134 - 2134209 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_030158.1 |
|
1 - 1 |
41 |
66 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
INS-IGF2 readthrough |
INS-IGF2 |
INSIGF |
|
Sense |
Chr. - 2129112 - 2161209 on Build GRCh38 |
Downstream |
IGF2 antisense RNA |
IGF2-AS |
IGF2-AS1, IGF2AS, PEG8 |
6.3kb
|
Anti-sense |
Chr. - 2140512 - 2148666 on Build GRCh38 |
View Full Product Details