Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMC10
Entrez Gene ID
83787
Gene Name
armadillo repeat containing 10
Gene Aliases
PNAS-112, PNAS112, PSEC0198, SVH
Chromosome Location
Chr.7: 103074727 - 103099763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161009.2 NP_001154481.1 1 - 2 530 99
NM_001161011.2 NP_001154483.1 1 - 2 530 99
NM_001161012.2 NP_001154484.1 1 - 2 530 99
NM_001161013.2 NP_001154485.1 1 - 2 530 99
GenBank mRNA AF275808.1 - 1 - 2 38 99
AK225082.1 - 1 - 2 263 99
AK291818.1 - 1 - 2 258 99
AK301565.1 - 1 - 2 263 99
AY150851.1 - 1 - 2 530 99
AY150853.1 - 1 - 2 530 99
BC003586.1 - 1 - 2 250 99

View Full Product Details