The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244755.1 | NP_001231684.1 | 2 - 2 | 319 | 82 |
NM_005752.4 | NP_005743.4 | 3 - 3 | 540 | 82 | |
GenBank mRNA | AA399390.1 | - | 2 - 2 | 320 | 82 |
AK313316.1 | - | 3 - 3 | 540 | 82 | |
AY358376.1 | - | 3 - 3 | 492 | 82 | |
BC104414.2 | - | 3 - 3 | 82 | ||
BC104415.2 | - | 3 - 3 | 529 | 82 | |
DA998903.1 | - | 3 - 3 | 540 | 82 |