Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CRIP2
- Entrez Gene ID
-
1397
- Gene Name
- cysteine rich protein 2
- Gene Aliases
-
CRIP, CRP2, ESP1
- Chromosome Location
-
Chr.14: 105472938 - 105480170 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073085.1 |
|
1 - 2 |
357 |
82 |
GenBank mRNA |
AK311402.1 |
- |
1 - 2 |
357 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB991533.1 |
- |
|
379 |
|
Chr. - - [-] on Build |
82 |
GenBankmRNA |
BX356167.2 |
- |
1 - 2 |
354 |
|
Chr. 14 - 105477249 - 105479839 [+] on Build GRCh38 |
82 |
GenBankmRNA |
DB190747.1 |
- |
1 - 2 |
357 |
|
Chr. 14 - 105477246 - 105478788 [+] on Build GRCh38 |
82 |
GenBankmRNA |
BX402264.2 |
- |
1 - 2 |
354 |
|
Chr. 14 - 105477249 - 105479631 [+] on Build GRCh38 |
82 |
GenBankmRNA |
DA667621.1 |
- |
1 - 2 |
357 |
|
Chr. 14 - 105477246 - 105478754 [+] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
metastasis associated 1 |
MTA1 |
|
-6.5kb
|
Sense |
Chr. - 105419849 - 105470729 on Build GRCh38 |
Downstream |
cysteine rich protein 1 |
CRIP1 |
CRHP, CRIP, CRP-1, CRP1 |
8.6kb
|
Sense |
Chr. - 105486920 - 105488789 on Build GRCh38 |
View Full Product Details