The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073199.1 | 3 - 4 | 357 | 70 | |
GenBank mRNA | AK000670.1 | - | 3 - 4 | 358 | 70 |
AK000701.1 | - | 2 - 3 | 2292 | 70 | |
BP382918.1 | - | 3 - 4 | 357 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BG959766.1 | - | 396 | Chr. - - [-] on Build | 70 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | LUC7 like 3 pre-mRNA splicing factor | LUC7L3 | CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A | -8.3kb | Anti-sense | Chr. - 50719565 - 50754269 on Build GRCh38 |
Downstream | microRNA 8059 | MIR8059 | hsa-mir-8059 | 5.1kb | Anti-sense | Chr. - 50768650 - 50768730 on Build GRCh38 |