The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024766.4 | NP_079042.1 | 2 - 3 | 418 | 106 |
XM_011533111.2 | XP_011531413.1 | 2 - 3 | 384 | 106 | |
XM_017004970.1 | XP_016860459.1 | 2 - 3 | 608 | 106 | |
XM_017004971.1 | XP_016860460.1 | 2 - 3 | 608 | 106 | |
XM_017004972.1 | XP_016860461.1 | 2 - 3 | 608 | 106 | |
XM_017004973.1 | XP_016860462.1 | 2 - 3 | 608 | 106 | |
XM_017004974.1 | XP_016860463.1 | 2 - 3 | 608 | 106 | |
XM_017004975.1 | XP_016860464.1 | 2 - 3 | 608 | 106 | |
XM_017004976.1 | XP_016860465.1 | 2 - 3 | 608 | 106 | |
XM_017004977.1 | XP_016860466.1 | 2 - 3 | 608 | 106 | |
XM_017004978.1 | XP_016860467.1 | 2 - 3 | 608 | 106 | |
XM_017004979.1 | XP_016860468.1 | 2 - 3 | 608 | 106 | |
XM_017004980.1 | XP_016860469.1 | 2 - 3 | 608 | 106 | |
XM_017004981.1 | XP_016860470.1 | 2 - 3 | 608 | 106 | |
GenBank mRNA | AK027104.1 | - | 2 - 3 | 358 | 106 |
BC029359.1 | - | 2 - 3 | 370 | 106 | |
BC053733.1 | - | 2 - 3 | 345 | 106 | |
DA760277.1 | - | 2 - 3 | 420 | 106 | |
DB461740.1 | - | 2 - 3 | 400 | 106 |