The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282534.1 | NP_001269463.1 | 1 - 2 | 343 | 86 |
NR_104210.1 | 1 - 2 | 343 | 86 | ||
XM_011517101.2 | XP_011515403.1 | 1 - 2 | 369 | 86 | |
XM_011517102.2 | XP_011515404.1 | 1 - 2 | 422 | 86 | |
GenBank mRNA | AF212829.1 | - | 1 - 2 | 279 | 86 |
AF248241.1 | - | 1 - 2 | 343 | 86 | |
AF257080.1 | - | 1 - 2 | 285 | 86 | |
AF279809.1 | - | 1 - 2 | 279 | 86 | |
AY190605.1 | - | 1 - 2 | 279 | 86 | |
BC075079.2 | - | 1 - 2 | 328 | 86 | |
BC075080.2 | - | 1 - 2 | 328 | 86 | |
BC112063.1 | - | 1 - 2 | 328 | 86 | |
BC112065.1 | - | 1 - 2 | 328 | 86 |