The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037225.2 | NP_001032302.1 | 9 - 10 | 821 | 69 |
NM_001300803.1 | NP_001287732.1 | 7 - 8 | 651 | 69 | |
NM_001318808.1 | NP_001305737.1 | 10 - 11 | 1599 | 69 | |
XM_005273918.2 | XP_005273975.1 | 10 - 11 | 879 | 69 | |
XM_006718502.2 | XP_006718565.1 | 10 - 11 | 996 | 69 | |
XM_006718503.3 | XP_006718566.1 | 10 - 11 | 882 | 69 | |
XM_006718504.2 | XP_006718567.1 | 9 - 10 | 932 | 69 | |
XM_006718505.2 | XP_006718568.1 | 7 - 8 | 762 | 69 | |
XM_011544940.2 | XP_011543242.1 | 10 - 11 | 877 | 69 | |
XM_011544941.2 | XP_011543243.1 | 4 - 5 | 578 | 69 | |
GenBank mRNA | AK093130.1 | - | 10 - 11 | 1593 | 69 |
BC033501.1 | - | 7 - 8 | 583 | 69 | |
BC106951.2 | - | 8 - 9 | 616 | 69 | |
BC106952.1 | - | 8 - 9 | 616 | 69 | |
LC068587.1 | - | 8 - 9 | 723 | 69 |