The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301873.1 | NP_001288802.1 | 1 - 2 | 142 | 87 |
NM_001301874.1 | NP_001288803.1 | 1 - 2 | 142 | 87 | |
NM_148672.3 | NP_683513.1 | 1 - 2 | 142 | 87 | |
GenBank mRNA | AF110384.1 | - | 1 - 2 | 121 | 87 |
AF220210.1 | - | 1 - 2 | 67 | 87 | |
AF266504.1 | - | 1 - 2 | 119 | 87 | |
AK312120.1 | - | 1 - 2 | 125 | 87 | |
BC062668.1 | - | 1 - 2 | 84 | 87 | |
BC069532.1 | - | 1 - 2 | 117 | 87 | |
BX486632.1 | - | 1 - 2 | 143 | 87 | |
HM067700.1 | - | 1 - 2 | 67 | 87 |