Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP57L1
Entrez Gene ID
285753
Gene Name
centrosomal protein 57 like 1
Gene Aliases
C6orf182, bA487F23.2, cep57R
Chromosome Location
Chr.6: 109095130 - 109167696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083535.1 NP_001077004.1 5 - 6 906 97
NM_001271852.1 NP_001258781.1 3 - 4 755 97
NM_001271853.1 NP_001258782.1 3 - 4 755 97
NM_173830.4 NP_776191.1 3 - 4 521 97
XM_005266935.4 XP_005266992.3 3 - 4 454 97
XM_005266936.4 XP_005266993.3 3 - 4 454 97
XM_005266938.4 XP_005266995.3 3 - 4 454 97
XM_006715462.3 XP_006715525.1 4 - 5 818 97
XM_011535759.2 XP_011534061.1 3 - 4 768 97
XM_011535763.2 XP_011534065.2 3 - 4 454 97
XM_017010764.1 XP_016866253.1 4 - 5 957 97
XM_017010765.1 XP_016866254.1 5 - 6 1212 97
XM_017010767.1 XP_016866256.1 3 - 4 768 97
XM_017010768.1 XP_016866257.1 3 - 4 582 97
XM_017010769.1 XP_016866258.1 3 - 4 521 97
XM_017010770.1 XP_016866259.1 5 - 6 1212 97
GenBank mRNA AK289910.1 - 3 - 4 387 97
BC033448.2 - 3 - 4 521 97
BC064365.1 - 3 - 4 750 97
BM787514.1 - 5 - 6 563 97
DB204503.1 - 1 - 2 127 97

View Full Product Details