Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN3
Entrez Gene ID
1070
Gene Name
centrin 3
Gene Aliases
CDC31, CEN3
Chromosome Location
Chr.5: 90393335 - 90409786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297768.1 NP_001284697.1 4 - 5 464 121
GenBank mRNA BU569574.1 - 5 - 6 413 121

View Full Product Details