Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WT1-AS
Entrez Gene ID
51352
Gene Name
WT1 antisense RNA
Gene Aliases
WIT-1, WIT1, WT1-AS1, WT1AS
Chromosome Location
Chr.11: 32435518 - 32458769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_120549.1 1 - 2 324 93


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Wilms tumor 1 WT1 AWT1, EWS-WT1, GUD, NPHS4, WAGR, WIT-2, WT33 Anti-sense Chr. - 32387775 - 32435535 on Build GRCh38
Downstream eukaryotic translation initiation factor 3 subunit M EIF3M B5, GA17, PCID1, TANGO7, hfl-B5 146.7kb Sense Chr. - 32583767 - 32602873 on Build GRCh38

View Full Product Details