Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101929023
- Entrez Gene ID
-
101929023
- Gene Name
- uncharacterized LOC101929023
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 116423724 - 116478826 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125973.1 |
|
3 - 4 |
302 |
120 |
GenBank mRNA |
AI672558.1 |
- |
1 - 2 |
|
120 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG054522.1 |
- |
1 - 2 |
|
|
Chr. 1 - 116453035 - 116478172 [+] on Build GRCh38 |
120 |
GenBankmRNA |
AI990641.1 |
- |
1 - 2 |
|
|
Chr. 1 - 116453035 - 116478826 [+] on Build GRCh38 |
120 |
GenBankmRNA |
AI240854.1 |
- |
1 - 2 |
|
|
Chr. 1 - 116453035 - 116478227 [+] on Build GRCh38 |
120 |
GenBankmRNA |
BF590025.1 |
- |
1 - 2 |
|
|
Chr. 1 - 116453034 - 116466442 [+] on Build GRCh38 |
120 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATP1A1 antisense RNA 1 |
ATP1A1-AS1 |
ATP1A1OS, C1orf203 |
-34.4kb
|
Sense |
Chr. - 116392865 - 116418622 on Build GRCh38 |
Downstream |
uncharacterized LOC105378922 |
LOC105378922 |
|
37.8kb
|
Sense |
Chr. - 116493016 - 116505088 on Build GRCh38 |
View Full Product Details