Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- STPG2-AS1
- Entrez Gene ID
-
101410545
- Gene Name
- STPG2 antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 97366926 - 97490164 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_102713.1 |
|
5 - 6 |
368 |
102 |
GenBank mRNA |
DA719663.1 |
- |
5 - 6 |
371 |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sperm tail PG-rich repeat containing 2 |
STPG2 |
C4orf37 |
|
Anti-sense |
Chr. - 97441450 - 98143473 on Build GRCh38 |
Downstream |
Rap1 GTPase-GDP dissociation stimulator 1 |
RAP1GDS1 |
GDS1, SmgGDS |
771.2kb
|
Sense |
Chr. - 98261376 - 98443861 on Build GRCh38 |
View Full Product Details