Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ITPKB-IT1
- Entrez Gene ID
-
100506443
- Gene Name
- ITPKB intronic transcript 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 226656080 - 226675068 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103784.1 |
|
1 - 2 |
148 |
113 |
GenBank mRNA |
BI906918.1 |
- |
1 - 2 |
147 |
113 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR744533.1 |
- |
1 - 2 |
45 |
|
Chr. 1 - 226656642 - 226674965 [-] on Build GRCh38 |
113 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
stum, mechanosensory transduction mediator homolog |
STUM |
C1orf95 |
-46.9kb
|
Anti-sense |
Chr. - 226548238 - 226609214 on Build GRCh38 |
Downstream |
presenilin 2 |
PSEN2 |
AD3L, AD4, CMD1V, PS2, STM2 |
195.5kb
|
Anti-sense |
Chr. - 226870572 - 226903829 on Build GRCh38 |
View Full Product Details