The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001565.3 | NP_001556.2 | 1 - 2 | 161 | 74 |
GenBank mRNA | BC010954.1 | - | 1 - 2 | 126 | 74 |