Product Details

Assay Reference Genome
Location

Chr.6:49457124 on build GRCh38
Cytoband
6p12.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
4594
Gene Symbol
MUT
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
Location
Chr.6:49430360-49463328 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MUT NM_000255.3 NP_000246.2
XM_005249143.3 XP_005249200.1
AK292568.1
AK312611.1
BC016282.1 AAH16282.1
BT007434.1 AAP36102.1
BX647789.1
KF030882.1
KU178201.1
KU178202.1
M65131.1 AAA59569.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5960n100 Chr6:49322472 - 49684411 on Build GRCh38 Gain CENPQ CRISP2 C6orf141 MUT RHAG GLYATL3
dgv10654n54 Chr6:49453528 - 49478796 on Build GRCh38 Gain CENPQ MUT

View Full Product Details