Product Details
- Assay Reference Genome
Location
Chr.7:92498666 on build GRCh38
- Cytoband
- 7q21.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs79758797] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 5189
- Gene Symbol
- PEX1
- Gene Name
- peroxisomal biogenesis factor 1
- Gene Aliases
- HMLR1, PBD1A, PBD1B, ZWS, ZWS1
- Location
- Chr.7:92487023-92528531 on build GRCh38
- Assay Gene Location
- Within Intron 16
View Full Product Details