Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
VPS35 | NM_018206.5 | NP_060676.2 | ||
XM_005256045.3 | XP_005256102.1 | |||
XM_011523227.2 | XP_011521529.1 | |||
AF175265.1 | AAF89953.1 | |||
AF186382.1 | AAG40619.1 | |||
AF191298.2 | AAF02778.2 | |||
AK001614.1 | BAA91790.1 | |||
AK223218.1 | BAD96938.1 | |||
AL136888.1 | CAB66822.1 | |||
BC010362.1 | AAH10362.1 | |||
BC093036.1 | AAH93036.1 | |||
DB049686.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv572536 | Chr16:46660737 - 46693473 on Build GRCh38 | Loss | ORC6 VPS35 |
nsv526656 | Chr16:46505480 - 46812751 on Build GRCh38 | Gain | SHCBP1 MYLK3 ORC6 ANKRD26P1 VPS35 C16orf87 |
dgv2975n100 | Chr16:46429858 - 46768065 on Build GRCh38 | Gain | SHCBP1 MYLK3 ORC6 ANKRD26P1 VPS35 |
esv3571142 | Chr16:46430577 - 46840772 on Build GRCh38 | Gain | SHCBP1 MYLK3 ORC6 ANKRD26P1 VPS35 C16orf87 |
esv3638540 | Chr16:46678001 - 46738780 on Build GRCh38 | Gain | MYLK3 ORC6 VPS35 |
esv3892855 | Chr16:46671431 - 46750308 on Build GRCh38 | Loss | MYLK3 ORC6 VPS35 |
esv3638534 | Chr16:46547097 - 46716009 on Build GRCh38 | Gain | SHCBP1 MYLK3 ORC6 ANKRD26P1 VPS35 |