Product Details

Assay Reference Genome
Location

Chr.1:7973327 on build GRCh38
Cytoband
1p36.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
11315
Gene Symbol
PARK7
Gene Name
Parkinsonism associated deglycase
Gene Aliases
DJ-1, DJ1, HEL-S-67p
Location
Chr.1:7961654-7985282 on build GRCh38
Assay Gene Location
Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PARK7 NM_001123377.1 NP_001116849.1
NM_007262.4 NP_009193.2
XM_005263424.3 XP_005263481.1
AB073864.1 BAB71782.1
AF021819.1 AAC12806.1
AK130886.1
AK312000.1
BC008188.2 AAH08188.1
BP248330.1
D61380.2 BAA09603.2
EU794641.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1521 Chr1:7954179 - 7987433 on Build GRCh38 Insertion PARK7

View Full Product Details