Product Details

Assay Reference Genome
Location

Chr.1:40301965 on build GRCh38
Cytoband
1p34.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1298
Gene Symbol
COL9A2
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
Location
Chr.1:40300487-40317653 on build GRCh38
Assay Gene Location
Within Intron 32
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
COL9A2 NM_001852.3 NP_001843.1
XM_006710365.3 XP_006710428.1
XM_011540715.2 XP_011539017.1
XM_011540716.2 XP_011539018.1
XM_011540717.2 XP_011539019.1
XM_017000332.1 XP_016855821.1
XM_017000333.1 XP_016855822.1
BC136326.1
BC136327.1
M95610.1 AAA80977.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv950687 Chr1:40294329 - 40321928 on Build GRCh38 Deletion COL9A2

View Full Product Details