Product Details

Assay Reference Genome
Location

Chr.8:63085469 on build GRCh38
Cytoband
8q12.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
7274
Gene Symbol
TTPA
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
Location
Chr.8:63058409-63086523 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TTPA NM_000370.3 NP_000361.1
XM_006716468.3 XP_006716531.1
BC058000.1 AAH58000.1
D49488.1 BAA08449.1
U21938.1 AAA64309.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv611448 Chr8:63038678 - 63167927 on Build GRCh38 Loss YTHDF3-AS1 GGH TTPA

View Full Product Details