Product Details

Assay Reference Genome
Location

Chr.6:43609587 on build GRCh38
Cytoband
6p21.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5429
Gene Symbol
POLH
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
Location
Chr.6:43576141-43620523 on build GRCh38
Assay Gene Location
Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
POLH NM_001291969.1 NP_001278898.1
NM_001291970.1 NP_001278899.1
NM_006502.2 NP_006493.1
XM_005249186.3 XP_005249243.1
XM_011514698.2 XP_011513000.1
AB024313.1 BAA81666.1
AF158185.1 AAD43810.1
AJ420484.1
AK023893.1
AK122807.1
AK294430.1
AK307660.1
AK316151.1
BC015742.1 AAH15742.1

View Full Product Details