Product Details

Assay Reference Genome
Location

Chr.19:16201027 on build GRCh38
Cytoband
19p13.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8907
Gene Symbol
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
Location
Chr.19:16197854-16235345 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
AP1M1 NM_001130524.1 NP_001123996.1
NM_032493.3 NP_115882.1
AB209808.1 BAD93045.1
AF290613.1 AAK28024.1
AK027528.1
AK293188.1
AK297824.1
BC017469.2 AAH17469.1
DC386056.1
DQ059565.1 AAY54246.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3643792 Chr19:16195112 - 16204140 on Build GRCh38 Gain AP1M1
nsv833767 Chr19:16121391 - 16347760 on Build GRCh38 Loss CIB3 KLF2 RAB8A HSH2D FAM32A AP1M1

View Full Product Details