Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KCNE3 | NM_005472.4 | NP_005463.1 | ||
XM_011544713.2 | XP_011543015.1 | |||
XM_017017047.1 | XP_016872536.1 | |||
XM_017017048.1 | XP_016872537.1 | |||
XM_017017050.1 | 1 | 1907 | XP_016872539.1 | |
XM_017017051.1 | XP_016872540.1 | |||
XM_017017052.1 | XP_016872541.1 | |||
CD709836.1 | ||||
CR936625.1 | CAI56768.1 | |||
DQ192291.1 | ABB00304.1 | |||
DR006135.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041154 | Chr11:74403756 - 74466864 on Build GRCh38 | Loss | KCNE3 |
esv34979 | Chr11:74167643 - 74822407 on Build GRCh38 | Gain | RNF169 LIPT2 CHRDL2 LOC100287896 MIR548AL PGM2L1 C2CD3 PPME1 KCNE3 POLD3 P4HA3 MIR4696 LOC101928580 |
esv2759837 | Chr11:74131564 - 74846552 on Build GRCh38 | Gain | XRRA1 RNF169 LIPT2 CHRDL2 LOC100287896 MIR548AL PGM2L1 C2CD3 PPME1 KCNE3 POLD3 P4HA3 MIR4696 LOC101928580 |
nsv555443 | Chr11:74398121 - 74469444 on Build GRCh38 | Loss | KCNE3 MIR548AL PGM2L1 |
nsv832209 | Chr11:74444838 - 74618074 on Build GRCh38 | Gain | KCNE3 POLD3 LIPT2 LOC100287896 |
dgv209e214 | Chr11:74225600 - 74828943 on Build GRCh38 | Gain | KCNE3 RNF169 POLD3 LIPT2 P4HA3 CHRDL2 MIR4696 LOC100287896 MIR548AL PGM2L1 LOC101928580 PPME1 |
nsv393 | Chr11:74424817 - 74470372 on Build GRCh38 | Deletion | KCNE3 |
nsv825994 | Chr11:74225622 - 74823410 on Build GRCh38 | Gain | KCNE3 RNF169 POLD3 LIPT2 P4HA3 CHRDL2 MIR4696 LOC100287896 MIR548AL PGM2L1 LOC101928580 PPME1 |