Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199274.1 | NP_001186203.1 | 5, 6 | 1068 | |
NM_134017.2 | NP_598778.1 | 5, 6 | 872 | ||
GenBank mRNA | AB070267.1 | - | 5, 6 | 839 | |
AK075600.1 | - | 5, 6 | 773 | ||
AK075947.1 | - | 5, 6 | 807 | ||
AK153126.1 | - | 5, 6 | 826 | ||
AK162087.1 | - | 5, 6 | 807 | ||
BC003457.1 | - | 5, 6 | 803 | 2648452 |