Gene Transcripts

Gene Symbol
Manba
Entrez Gene ID
110173
Gene Name
mannosidase, beta A, lysosomal
Gene Aliases
2410030O07Rik, B930014J03Rik, Bmn
Chromosome Location
Chr. 3: 135485575 - 135571404 on Build GRCm38
UniGene ID
Mm.280536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027288.3 NP_081564.3 9, 10 1303
GenBank mRNA AF306557.1 - 9, 10 1307
AK036505.1 - 10, 11 1403
AK047057.1 - 9, 10 1296
AK142165.1 - 9, 10 1268
AK154172.1 - 9, 10 1285
AK166022.1 - 9, 10 1286
AK170290.1 - 9, 10 1300
BC031409.1 - 9, 10 1246 4952710

View Full Product Details