Gene Transcripts

Gene Symbol
Chrna1
Entrez Gene ID
11435
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle)
Gene Aliases
AI385656, AI608266, Achr-1, Acra
Chromosome Location
Chr. 2: 73563281 - 73580338 on Build GRCm38
UniGene ID
Mm.4583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007389.5 NP_031415.2 5 410
GenBank mRNA AK029177.1 - 5 379
AK132589.1 - 5 350
BC125439.1 - 5 410 40129954
BC137678.1 - 5 410 8860697
X03986.1 - 5 367

View Full Product Details