Gene Transcripts
- Gene Symbol
- Bmp1
- Entrez Gene ID
- 12153
- Gene Name
- bone morphogenetic protein 1
- Gene Aliases
- Pcp, Tld
- Chromosome Location
- Chr. 14: 70474555 - 70520712 on Build GRCm38
- UniGene ID
- Mm.27757
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009755.3 |
NP_033885.2
|
6, 7 |
1085 |
|
GenBank mRNA |
AK004995.2 |
- |
6, 7 |
720 |
|
AK147534.1 |
- |
4, 5 |
488 |
|
AK154239.1 |
- |
6, 7 |
1087 |
|
AK158218.1 |
- |
6, 7 |
1078 |
|
AK220295.1 |
- |
6, 7 |
1096 |
|
BC053923.1 |
- |
6, 7 |
1012 |
|
BC063079.1 |
- |
2, 3 |
215 |
|
BC066062.1 |
- |
6, 7 |
1013 |
6849066 |
L24755.1 |
- |
6, 7 |
862 |
|
View Full Product Details