Gene Transcripts

Gene Symbol
Eya4
Entrez Gene ID
14051
Gene Name
eyes absent 4 homolog (Drosophila)
Gene Aliases
B130023L16Rik
Chromosome Location
Chr. 10: 23102962 - 23350856 on Build GRCm38
UniGene ID
Mm.332128
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010167.4 NP_034297.2 15 1573
GenBank mRNA AK041625.1 - 15 1574
BC120898.1 - 15 1557 40061597
Y17115.1 - 16 1684

View Full Product Details