Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010476.3 | NP_034606.3 | 5, 6 | 709 | |
GenBank mRNA | AK028380.1 | - | 5, 6 | 665 | |
AK050211.1 | - | 5, 6 | 666 | ||
AK077914.1 | - | 5, 6 | 707 | ||
BC011464.1 | - | 5, 6 | 673 | ||
Y15733.1 | - | 5, 6 | 699 |