Gene Transcripts

Gene Symbol
Lamb3
Entrez Gene ID
16780
Gene Name
laminin, beta 3
Gene Aliases
Chromosome Location
Chr. 1: 193294353 - 193343878 on Build GRCm38
UniGene ID
Mm.435441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277928.1 NP_001264857.1 16 2454
NM_008484.2 NP_032510.2 16 2493
GenBank mRNA AK155769.1 - 3306
BC008516.1 - 16 2454 3592888
CX237661.1 - 2 356
U43298.1 - 16 2469

View Full Product Details