Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_130878.2 | NP_570948.1 | 14, 15 | 1719 | |
GenBank mRNA | AB093300.1 | - | 14, 15 | 2135 | |
AF426393.1 | - | 14, 15 | 1606 | ||
AK143151.1 | - | 14, 15 | 1660 | ||
BC042454.1 | - | 14, 15 | 1629 | 5369576 | |
BC058270.1 | - | 14, 15 | 1719 | 6490597 |