Gene Transcripts

Gene Symbol
Clec4d
Entrez Gene ID
17474
Gene Name
C-type lectin domain family 4, member d
Gene Aliases
Clecsf8, Mpcl, mcl
Chromosome Location
Chr. 6: 123262107 - 123275268 on Build GRCm38
UniGene ID
Mm.441101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163161.1 NP_001156633.1 4 505
NM_010819.4 NP_034949.3 4 508
GenBank mRNA AF061272.1 - 4 440
AK089500.1 - 4 499
AK150926.1 - 4 411
AK171409.1 - 4 446
BC099471.1 - 4 418 30845162

View Full Product Details