Gene Transcripts

Gene Symbol
Slc3a1
Entrez Gene ID
20532
Gene Name
solute carrier family 3, member 1
Gene Aliases
D2H, NBAT, NTAA, RBAT
Chromosome Location
Chr. 17: 85028347 - 85064241 on Build GRCm38
UniGene ID
Mm.227176
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009205.2 NP_033231.2 5 1001
GenBank mRNA AK052623.1 - 5 972
AK132275.1 - 5 973
BC013441.1 - 5 943 4222972
D88533.1 - 5 961

View Full Product Details