Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009206.2 | NP_033232.2 | 7 | 1400 | |
GenBank mRNA | AF035526.1 | - | 8 | 1283 | |
AK138398.1 | - | 7 | 1355 | ||
AK147161.1 | - | 7 | 738 | ||
BC072588.1 | - | 7 | 1383 | ||
BC090964.1 | - | 7 | 952 | 30625511 |