Gene Transcripts

Gene Symbol
Asxl3
Entrez Gene ID
211961
Gene Name
additional sex combs like 3 (Drosophila)
Gene Aliases
C230079D11Rik, D430002O22Rik, D930044O18Rik, Gm329, Gm945
Chromosome Location
Chr. 18: 22344077 - 22530227 on Build GRCm38
UniGene ID
Mm.392310
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167777.1 NP_001161249.1 13 2925
GenBank mRNA AK082659.1 - 13 2925

View Full Product Details