Gene Transcripts

Gene Symbol
Ctage5
Entrez Gene ID
217615
Gene Name
CTAGE family, member 5
Gene Aliases
D12Bwg0579e, Mea6, Mgea, Mgea6
Chromosome Location
Chr. 12: 59129740 - 59190220 on Build GRCm38
UniGene ID
Mm.244118
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146034.3 NP_666146.3 5, 6 473
GenBank mRNA AK087188.1 - 4, 5 388
AK208297.1 - 5, 6 346
BC026864.1 - 5, 6 425 2647214

View Full Product Details