Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011620.3 | NP_035750.2 | 6, 7 | 164 | |
GenBank mRNA | AA754928.1 | - | 6, 7 | 141 | |
DV049834.1 | - | 6, 7 | 211 | ||
L48990.1 | - | 6, 7 | 100 | ||
L48991.1 | - | 3, 4 | 82 | ||
L49018.1 | - | 3, 4 | 82 | ||
U77779.1 | - | 6, 7 | 157 |