Gene Transcripts

Gene Symbol
Vmn1r51
Entrez Gene ID
22296
Gene Name
vomeronasal 1 receptor 51
Gene Aliases
V1R, V1r1, V1ra1, VN12, mV1R1
Chromosome Location
Chr. 6: 90122643 - 90130645 on Build GRCm38
UniGene ID
Mm.20471
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011683.2 NP_035813.2 3 1697
GenBank mRNA BC046308.1 - 3 1697
Y12725.1 - 2 1667

View Full Product Details