Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153791.2 | NP_722486.2 | 6 | 1811 | |
GenBank mRNA | AK034869.1 | - | 5 | 1405 | |
AK052314.1 | - | 4 | 1023 | ||
AK129391.1 | - | 4 | 969 | ||
AK168389.1 | - | 6 | 1746 | ||
AK181512.1 | - | 2 | 215 | ||
BC025645.1 | - | 2 | 196 | ||
BC038031.1 | - | 5 | 2103 | 4021754 |