Gene Transcripts
- Gene Symbol
- Fhod3
- Entrez Gene ID
- 225288
- Gene Name
- formin homology 2 domain containing 3
- Gene Aliases
- A930009H06Rik, FHOS2, mKIAA1695
- Chromosome Location
- Chr. 18: 24708456 - 25133507 on Build GRCm38
- UniGene ID
- Mm.329322
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289654.1 |
NP_001276583.1
|
21, 22 |
4647 |
|
NM_001289655.1 |
NP_001276584.1
|
22, 23 |
5004 |
|
NM_175276.4 |
NP_780485.2
|
23, 24 |
5100 |
|
GenBank mRNA |
AB078608.1 |
- |
23, 24 |
4278 |
|
AB078609.1 |
- |
21, 22 |
3825 |
|
AB100088.1 |
- |
22, 23 |
4182 |
|
AK148245.1 |
- |
21, 22 |
3789 |
|
AK220413.1 |
- |
19, 20 |
3264 |
|
BC060607.1 |
- |
8, 9 |
2056 |
|
BC138046.1 |
- |
21, 22 |
3747 |
8861065 |
View Full Product Details