Gene Transcripts
- Gene Symbol
- Heatr3
- Entrez Gene ID
- 234549
- Gene Name
- HEAT repeat containing 3
- Gene Aliases
- C030036P15Rik
- Chromosome Location
- Chr. 8: 88137885 - 88171952 on Build GRCm38
- UniGene ID
- Mm.36291
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172757.3 |
NP_766345.3
|
5, 6 |
751 |
|
GenBank mRNA |
AK047782.1 |
- |
5, 6 |
782 |
|
AK052635.1 |
- |
5, 6 |
778 |
|
AK134510.1 |
- |
5, 6 |
756 |
|
AK136424.1 |
- |
5, 6 |
761 |
|
AK140522.1 |
- |
5, 6 |
769 |
|
AK163715.1 |
- |
5, 6 |
738 |
|
AK165541.1 |
- |
5, 6 |
645 |
|
BC030373.1 |
- |
1, 2 |
96 |
|
BC065060.1 |
- |
5, 6 |
751 |
6402217 |
View Full Product Details