Gene Transcripts

Gene Symbol
Sec31b
Entrez Gene ID
240667
Gene Name
Sec31 homolog B (S. cerevisiae)
Gene Aliases
Gm341, Sec31l2
Chromosome Location
Chr. 19: 44516957 - 44545848 on Build GRCm38
UniGene ID
Mm.329873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033343.1 NP_001028515.1 8, 9 970
GenBank mRNA AK158017.1 - 7, 8 847

View Full Product Details