Gene Transcripts
- Gene Symbol
- Zc4h2
- Entrez Gene ID
- 245522
- Gene Name
- zinc finger, C4H2 domain containing
- Gene Aliases
- Gm372, mKIAA1166
- Chromosome Location
- Chr. X: 95639190 - 95658509 on Build GRCm38
- UniGene ID
- Mm.336158
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003916.2 |
NP_001003916.1
|
4, 5 |
764 |
|
NM_001289696.1 |
NP_001276625.1
|
4, 5 |
796 |
|
NM_001289697.1 |
NP_001276626.1
|
4, 5 |
743 |
|
GenBank mRNA |
AK129302.1 |
- |
4, 5 |
617 |
|
AK155445.1 |
- |
4, 5 |
764 |
|
AK162005.1 |
- |
4, 5 |
661 |
|
BC050850.1 |
- |
4, 5 |
694 |
6508566 |
BC079862.1 |
- |
4, 5 |
687 |
6807437 |
BC094942.1 |
- |
4, 5 |
705 |
6435125 |
BC119285.1 |
- |
4, 5 |
678 |
8734034 |
BC119311.1 |
- |
4, 5 |
679 |
8734060 |
View Full Product Details