Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013891.4 | NP_038919.1 | 6 | 1388 | |
GenBank mRNA | AB019436.1 | - | 6 | 1324 | |
AK078863.1 | - | 6 | 1389 | ||
AV370994.2 | - | 2 | 246 | ||
BC012648.1 | - | 6 | 1389 | 4218741 |