Gene Transcripts

Gene Symbol
Prss34
Entrez Gene ID
328780
Gene Name
protease, serine 34
Gene Aliases
C130091P12, Mcp11, mMcp-11
Chromosome Location
Chr. 17: 25298394 - 25300168 on Build GRCm38
UniGene ID
Mm.127166
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178372.2 NP_848459.1 3 427
GenBank mRNA AK081986.1 - 3 429
AY261775.1 - 3 427
BC109366.2 - 3 401

View Full Product Details