Gene Transcripts

Gene Symbol
Skor2
Entrez Gene ID
664805
Gene Name
SKI family transcriptional corepressor 2
Gene Aliases
Corl2, EG664805, Fussel18, Gm7348
Chromosome Location
Chr. 18: 76846734 - 76901032 on Build GRCm38
UniGene ID
Mm.329543
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109743.1 NP_001103213.1 8 3062
GenBank mRNA AB358976.1 - 8 3062

View Full Product Details