Gene Transcripts

Gene Symbol
Dfnb59
Entrez Gene ID
381375
Gene Name
deafness, autosomal recessive 59 (human)
Gene Aliases
Gm1001, Pjvk
Chromosome Location
Chr. 2: 76648327 - 76658554 on Build GRCm38
UniGene ID
Mm.330312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080711.2 NP_001074180.1 1 317
GenBank mRNA DQ365828.1 - 1 317

View Full Product Details